4-(1H-1,2,3-Triazol-1-yl)benzoic acid 98%

Supplier: Ambeed
Warning

A1027612-1G A1027612-250MG A1027612-5G
76671-200EA 81.69 USD
76671-200 76671-202 76671-204
4-(1H-1,2,3-Triazol-1-yl)benzoic acid 98%
4-(1H-1,2,3-Triazol-1-yl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₇N₃O₂
MW: 189.1708 g/mol
MDL Number: MFCD20483616
CAS Number: 216959-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR