N-(2-(Allyloxy)benzyl)-N-(benzo[d][1,3]dioxol-5-yl)-2-chloroacetamide 95%

Supplier: Ambeed
Danger

A1027513-1G A1027513-250MG A1027513-100MG A1027513-5G
76671-198EA 55 USD
76671-198 77619-690 77782-887 77782-888
N-(2-(Allyloxy)benzyl)-N-(benzo[d][1,3]dioxol-5-yl)-2-chloroacetamide 95%
N-(2-(Allyloxy)benzyl)-N-(benzo[d][1,3]dioxol-5-yl)-2-chloroacetamide

Precaución: for Research Use Only

Formula: C₁₉H₁₈ClNO₄
MDL Number: MFCD06379302
CAS Number: 851452-58-9
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR