4-Chloro-2-(4,5-dihydro-1,3-oxazol-2-yl)pyridine 95%

Supplier: Ambeed
Warning

A1027054-100MG A1027054-1G A1027054-250MG
76671-186EA 93.36 USD
76671-186 76671-188 76671-190
4-Chloro-2-(4,5-dihydro-1,3-oxazol-2-yl)pyridine 95%
4-Chloro-2-(4,5-dihydro-1,3-oxazol-2-yl)pyridine

Precaución: for Research Use Only

Formula: C₈H₇ClN₂O
MW: 182.6070 g/mol
MDL Number: MFCD22628379
CAS Number: 1423027-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR