2-(5-Nitro-1H-indol-3-yl)acetic acid 95%

Supplier: Ambeed
Warning

A1027044-100MG A1027044-1G A1027044-250MG
76671-180EA 24 USD
76671-180 76671-182 76671-184
2-(5-Nitro-1H-indol-3-yl)acetic acid 95%
2-(5-Nitro-1H-indol-3-yl)acetic acid

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O₄
MDL Number: MFCD16737545
CAS Number: 79473-05-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR