2-Hydroxy-3-(3-methoxyphenyl)propanoic acid 95%

Supplier: Ambeed
Warning

A1026522-100MG A1026522-1G A1026522-250MG
76671-172EA 59.42 USD
76671-172 76671-174 76671-176
2-Hydroxy-3-(3-methoxyphenyl)propanoic acid 95%
2-Hydroxy-3-(3-methoxyphenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₄
MW: 196.1999 g/mol
MDL Number: MFCD11935533
CAS Number: 1181574-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR