2-[(2,4-Difluorophenyl)amino]ethan-1-ol 95%

Supplier: Ambeed
Warning

A1026264-100MG A1026264-1G A1026264-250MG A1026264-5G
76671-164EA 25.47 USD
76671-164 76671-166 76671-168 76671-170
2-[(2,4-Difluorophenyl)amino]ethan-1-ol 95%
2-[(2,4-Difluorophenyl)amino]ethan-1-ol

Precaución: for Research Use Only

Formula: C₈H₉F₂NO
MW: 173.1600 g/mol
MDL Number: MFCD11153579
CAS Number: 1020993-12-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR