1,2-Bis(2-chloroethyl)disulfane 95%

Supplier: Ambeed
Warning

A1025991-100MG A1025991-250MG
76671-156EA 188 USD
76671-156 77619-684
1,2-Bis(2-chloroethyl)disulfane 95%
1,2-Bis(2-chloroethyl)disulfane

Precaución: for Research Use Only

Formula: C₄H₈Cl₂S₂
MW: 191.1423 g/mol
MDL Number: MFCD20621166
CAS Number: 1002-41-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR