(3R)-3-amino-3-(4-bromo-2-methylphenyl)propan-1-ol 95%

Supplier: Ambeed
Warning

A1025708-100MG A1025708-250MG
76671-150EA 129.43 USD
76671-150 76671-152
(3R)-3-amino-3-(4-bromo-2-methylphenyl)propan-1-ol 95%
(3R)-3-amino-3-(4-bromo-2-methylphenyl)propan-1-ol

Precaución: for Research Use Only

Formula: C₁₀H₁₄BrNO
MW: 244.1283 g/mol
MDL Number: MFCD18686343
CAS Number: 1270175-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR