2-(4-(Difluoromethyl)phenyl)acetic acid 95%

Supplier: Ambeed
Warning

A1025435-1G A1025435-100MG
76671-148EA 567 USD
76671-148 77782-881
2-(4-(Difluoromethyl)phenyl)acetic acid 95%
2-(4-(Difluoromethyl)phenyl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₈F₂O₂
MDL Number: MFCD09925293
CAS Number: 1000524-74-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR