1-(Ethenesulfonyl)-4-fluorobenzene 98%

Supplier: Ambeed
Danger

A1025218-1G A1025218-250MG
76671-138EA 233 USD
76671-138 76671-140
1-(Ethenesulfonyl)-4-fluorobenzene 98%
1-(Ethenesulfonyl)-4-fluorobenzene

Precaución: for Research Use Only

Formula: C₈H₇FO₂S
MDL Number: MFCD00540721
CAS Number: 28122-14-7
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR