(1S)-1-(4-Methylphenyl)ethan-1-ol 98%

Supplier: Ambeed
Warning

A1020323-1G A1020323-250MG A1020323-5G
76671-114EA 45.62 USD
76671-114 76671-116 76671-118
(1S)-1-(4-Methylphenyl)ethan-1-ol 98%
(1S)-1-(4-Methylphenyl)ethan-1-ol

Precaución: for Research Use Only

Formula: C₉H₁₂O
MW: 136.1910 g/mol
MDL Number: MFCD11046060
CAS Number: 51154-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR