Methyl 5-[4-(trifluoromethyl)phenyl]oxolane-2-carboxylate 95%

Supplier: Ambeed
Warning

A1014801-100MG A1014801-1G A1014801-250MG
76671-086EA 9 USD
76671-086 76671-088 76671-090
Methyl 5-[4-(trifluoromethyl)phenyl]oxolane-2-carboxylate 95%
Methyl 5-[4-(trifluoromethyl)phenyl]oxolane-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₃F₃O₃
MW: 274.2357 g/mol
MDL Number: MFCD22392037
CAS Number: 1421601-10-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR