4-Chloro-2-(chloromethyl)-1-methyl-benzene 97%

Supplier: Ambeed
Danger

A1014504-1G A1014504-250MG
76671-082EA 164.44 USD
76671-082 76671-084
4-Chloro-2-(chloromethyl)-1-methyl-benzene 97%
4-Chloro-2-(chloromethyl)-1-methyl-benzene

Precaución: for Research Use Only

Formula: C₈H₈Cl₂
MW: 175.0551 g/mol
MDL Number: MFCD12547163
CAS Number: 34060-72-5
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR