2-{1-[(tert-butoxy)carbonyl]-2,3-dihydro-1H-indol-3-yl}acetic acid 95%

Supplier: Ambeed
Warning

A1013370-100MG A1013370-1G
76671-072EA 39.26 USD
76671-072 76671-074
2-{1-[(tert-butoxy)carbonyl]-2,3-dihydro-1H-indol-3-yl}acetic acid 95%
2-{1-[(tert-butoxy)carbonyl]-2,3-dihydro-1H-indol-3-yl}acetic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₉NO₄
MW: 277.3157 g/mol
MDL Number: MFCD21983583
CAS Number: 1321843-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR