Ethyl 5-amino-1,2-dimethyl-1H-imidazole-4-carboxylate 95%

Supplier: Ambeed
Warning

A1013276-100MG A1013276-1G
76671-062EA 65.78 USD
76671-062 76671-064
Ethyl 5-amino-1,2-dimethyl-1H-imidazole-4-carboxylate 95%
Ethyl 5-amino-1,2-dimethyl-1H-imidazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₈H₁₃N₃O₂
MW: 183.2077 g/mol
MDL Number: MFCD16112515
CAS Number: 1249354-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR