N,3-Dimethyl-4-nitroaniline 95%

Supplier: Ambeed
Warning

A1012727-1G A1012727-250MG
76671-058EA 360.71 USD
76671-058 76671-060
N,3-Dimethyl-4-nitroaniline 95%
N,3-Dimethyl-4-nitroaniline

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
MW: 166.1772 g/mol
MDL Number: MFCD12169124
CAS Number: 52177-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR