5,6-Difluoro-1H-1,3-benzodiazol-2-amine 95%

Supplier: Ambeed
Warning

A1012400-100MG A1012400-1G A1012400-250MG
76671-052EA 58.35 USD
76671-052 76671-054 76671-056
5,6-Difluoro-1H-1,3-benzodiazol-2-amine 95%
5,6-Difluoro-1H-1,3-benzodiazol-2-amine

Precaución: for Research Use Only

Formula: C₇H₅F₂N₃
MW: 169.1315 g/mol
MDL Number: MFCD16845540
CAS Number: 142356-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR