3-Bromo-5-cyanobenzene-1-sulfonyl chloride 95%

Supplier: Ambeed
Danger

A1012232-250MG A1012232-50MG
76671-048EA 274.78 USD
76671-048 76671-050
3-Bromo-5-cyanobenzene-1-sulfonyl chloride 95%
3-Bromo-5-cyanobenzene-1-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₇H₃BrClNO₂S
MW: 280.53 g/mol
MDL Number: MFCD10697739
CAS Number: 49674-17-1
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR