5-Fluoro-2-methyl-6-nitro-1,3-benzoxazole 95%

Supplier: Ambeed
Warning

A1011856-100MG A1011856-250MG
76671-042EA 76.39 USD
76671-042 76671-044
5-Fluoro-2-methyl-6-nitro-1,3-benzoxazole 95%
5-Fluoro-2-methyl-6-nitro-1,3-benzoxazole

Precaución: for Research Use Only

Formula: C₈H₅FN₂O₃
MW: 196.1353 g/mol
MDL Number: MFCD22196495
CAS Number: 40703-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR