[2-(2-Methoxyethoxy)phenyl]methanol 95%

Supplier: Ambeed
Warning

A1011842-5G A1011842-1G A1011842-250MG
76671-040EA 518 USD
76671-040 77782-846 77782-847
[2-(2-Methoxyethoxy)phenyl]methanol 95%
[2-(2-Methoxyethoxy)phenyl]methanol

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₃
MDL Number: MFCD11149625
CAS Number: 1020929-44-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR