4-[(2,4-Dimethylphenyl)sulfanyl]aniline hydrochloride 95%

Supplier: Ambeed
Warning

A1009374-100MG A1009374-1G
76671-030EA 26.53 USD
76671-030 76671-032
4-[(2,4-Dimethylphenyl)sulfanyl]aniline hydrochloride 95%
4-[(2,4-Dimethylphenyl)sulfanyl]aniline hydrochloride

Precaución: for Research Use Only

Formula: C₁₄H₁₆ClNS
MW: 265.8015 g/mol
MDL Number: MFCD07284384
CAS Number: 1049755-55-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR