2-Chloro-N-(2,4,6-triiodophenyl)acetamide 95%

Supplier: Ambeed
Danger

A1007525-1G A1007525-250MG
76671-028EA 233 USD
76671-028 77782-825
2-Chloro-N-(2,4,6-triiodophenyl)acetamide 95%
2-Chloro-N-(2,4,6-triiodophenyl)acetamide

Precaución: for Research Use Only

Formula: C₈H₅ClI₃NO
MDL Number: MFCD02725651
CAS Number: 380346-57-6
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR