1-Benzyl-3-methyl-1H-imidazol-3-ium bromide 98%

Supplier: Ambeed
Warning

A1006843-1G A1006843-25G A1006843-5G
76671-022EA 10.61 USD
76671-022 76671-024 76671-026
1-Benzyl-3-methyl-1H-imidazol-3-ium bromide 98%
1-Benzyl-3-methyl-1H-imidazol-3-ium bromide

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrN₂
MW: 253.1383 g/mol
MDL Number: MFCD22375980
CAS Number: 65039-11-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR