N-(3-(2-(2-(3-Aminopropoxy)ethoxy)ethoxy)propyl)-5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamide 97%

Supplier: Ambeed
Warning

Synonyms: Biotin-PEG3-C3-NH2

A1006576-100MG A1006576-250MG A1006576-1G A1006576-50MG
76671-020EA 162.32 USD
76671-020 77595-694 77619-644 77619-646
N-(3-(2-(2-(3-Aminopropoxy)ethoxy)ethoxy)propyl)-5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamide 97%
N-(3-(2-(2-(3-Aminopropoxy)ethoxy)ethoxy)propyl)-5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamide

Precaución: for Research Use Only

Formula: C₂₀H₃₈N₄O₅S
MW: 446.6045 g/mol
MDL Number: MFCD28053681
CAS Number: 183896-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR