(11bR)-1,1,1-Trifluoro-N-(4-oxido-2,6-diphenyl-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)methanesulfonamide 97%
Supplier: Ambeed
|
Warning
|
A1006126-100MG
A1006126-1G
A1006126-250MG
A1006126-50MG
76670-994EA
81
USD
76670-994
76670-996
76670-998
76671-000
(11bR)-1,1,1-Trifluoro-N-(4-oxido-2,6-diphenyl-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)methanesulfonamide 97%
(11bR)-1,1,1-Trifluoro-N-(4-oxido-2,6-diphenyl-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)methanesulfonamide
Precaución: for Research Use Only
Formula:
C₃₃H₂₉F₃NO₅PS |
MDL Number:
MFCD33021017 CAS Number: 1261302-62-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...