(11aS)-N,N-Diisopropyl-4,5,6,7-tetrahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-12-amine 97%

Supplier: Ambeed
Warning

A1006114-100MG A1006114-1G A1006114-250MG
76670-986EA 107 USD
76670-986 76670-988 76670-990
(11aS)-N,N-Diisopropyl-4,5,6,7-tetrahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-12-amine 97%
(11aS)-N,N-Diisopropyl-4,5,6,7-tetrahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-12-amine

Precaución: for Research Use Only

Formula: C₂₃H₂₈NO₂P
MW: 381.45 g/mol
CAS Number: 477559-83-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR