(11BR)-N-(2,6-di(naphthalen-1-yl)-4-oxidodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulfonamide 97%

Supplier: Ambeed
Warning

A1006064-100MG A1006064-1G A1006064-250MG A1006064-50MG
76670-960EA 205 USD
76670-960 76670-962 76670-964 76670-966
(11BR)-N-(2,6-di(naphthalen-1-yl)-4-oxidodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulfonamide 97%
(11BR)-N-(2,6-di(naphthalen-1-yl)-4-oxidodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulfonamide

Precaución: for Research Use Only

Formula: C₄₁H₂₅F₃NO₅PS
CAS Number: 1010800-00-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR