(2R,3R)-3-(tert-Butyl)-4-(2,6-dimethoxyphenyl)-2-methyl-2,3-dihydrobenzo[d][1,3]oxaphosphole 97%

Supplier: Ambeed
Warning

A1005935-100MG A1005935-250MG
76670-926EA 171.87 USD
76670-926 77619-584
(2R,3R)-3-(tert-Butyl)-4-(2,6-dimethoxyphenyl)-2-methyl-2,3-dihydrobenzo[d][1,3]oxaphosphole 97%
(2R,3R)-3-(tert-Butyl)-4-(2,6-dimethoxyphenyl)-2-methyl-2,3-dihydrobenzo[d][1,3]oxaphosphole

Precaución: for Research Use Only

Formula: C₂₀H₂₅O₃P
MW: 344.3800 g/mol
MDL Number: MFCD31630763
CAS Number: 1477517-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR