2-(Hydroxymethyl)-4-methoxy-3,5-dimethylpyridine 1-oxide 97%

Supplier: Ambeed
Warning

A1005492-100MG A1005492-1G A1005492-250MG
76670-916EA 23 USD
76670-916 76670-918 76670-920
2-(Hydroxymethyl)-4-methoxy-3,5-dimethylpyridine 1-oxide 97%
2-(Hydroxymethyl)-4-methoxy-3,5-dimethylpyridine 1-oxide

Precaución: for Research Use Only

Formula: C₉H₁₃NO₃
MDL Number: MFCD27948804
CAS Number: 287118-45-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR