N-(4-Chloro-3-(trifluoromethyl)phenyl)-2-(2-(dimethylamino)ethoxy)-6-ethoxybenzamide 97%

Supplier: Ambeed
Warning

Synonyms: YF-2

A1005340-10MG A1005340-1MG A1005340-5MG A1005340-50MG
76670-910EA 37 USD
76670-910 76670-912 76670-914 77296-740
N-(4-Chloro-3-(trifluoromethyl)phenyl)-2-(2-(dimethylamino)ethoxy)-6-ethoxybenzamide 97%
N-(4-Chloro-3-(trifluoromethyl)phenyl)-2-(2-(dimethylamino)ethoxy)-6-ethoxybenzamide

Precaución: for Research Use Only

Formula: C₂₀H₂₂ClF₃N₂O₃
MW: 430.85 g/mol
MDL Number: MFCD22665763
CAS Number: 1311423-89-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR