1-(tert-Butyl)-3-(2,6-diisopropyl-4-phenoxyphenyl)urea 97%

Supplier: Ambeed
Warning

A1005328-100MG A1005328-1G A1005328-250MG
76670-904EA 53.05 USD
76670-904 76670-906 76670-908
1-(tert-Butyl)-3-(2,6-diisopropyl-4-phenoxyphenyl)urea 97%
1-(tert-Butyl)-3-(2,6-diisopropyl-4-phenoxyphenyl)urea

Precaución: for Research Use Only

Formula: C₂₃H₃₂N₂O₂
MW: 368.512 g/mol
MDL Number: MFCD03427402
CAS Number: 136337-67-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR