2-(3-(9,9-Dimethyl-9H-fluoren-2-yl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%

Supplier: Ambeed
Warning

A1005094-1G A1005094-5G A1005094-250MG
76670-902EA 10 USD
76670-902 77782-764 77782-763
2-(3-(9,9-Dimethyl-9H-fluoren-2-yl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%
2-(3-(9,9-Dimethyl-9H-fluoren-2-yl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₂₇H₂₉BO₂
CAS Number: 1005771-03-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR