7-(4-(9-Phenyl-9H-carbazol-2-yl)quinazolin-2-yl)-7H-dibenzo[c,g]carbazole 98+%

Supplier: Ambeed
Warning

A1005086-100MG A1005086-1G A1005086-250MG
76670-896EA 15.92 USD
76670-896 76670-898 76670-900
7-(4-(9-Phenyl-9H-carbazol-2-yl)quinazolin-2-yl)-7H-dibenzo[c,g]carbazole 98+%
7-(4-(9-Phenyl-9H-carbazol-2-yl)quinazolin-2-yl)-7H-dibenzo[c,g]carbazole

Precaución: for Research Use Only

Formula: C₄₆H₂₈N₄
MW: 636.7413 g/mol
CAS Number: 1831056-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR