(2S,3R,4R,5S,6R)-6-(Hydroxymethyl)-5-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2,3,4-triol 98% DNase-, RNase-free

Supplier: Ambeed
Warning

Synonyms: α-Lactose

A1004906-1G A1004906-5G A1004906-25G A1004906-100G A1004906-500G
76670-892EA 5.31 USD
76670-892 76670-894 77595-656 77619-538 77619-540
(2S,3R,4R,5S,6R)-6-(Hydroxymethyl)-5-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2,3,4-triol 98% DNase-, RNase-free
(2S,3R,4R,5S,6R)-6-(Hydroxymethyl)-5-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2,3,4-triol

Precaución: for Research Use Only

Formula: C₁₂H₂₂O₁₁
MW: 342.2965 g/mol
MDL Number: MFCD00064074
CAS Number: 14641-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR