Dimethyl mesitylboronate 98%

Supplier: Ambeed
Warning

A1004775-1G A1004775-5G A1004775-250MG
76670-878EA 85 USD
76670-878 76670-880 77782-741
Dimethyl mesitylboronate 98%
Dimethyl mesitylboronate

Precaución: for Research Use Only

Formula: C₁₁H₁₇BO₂
MW: 192.0625 g/mol
MDL Number: MFCD32068248
CAS Number: 34907-53-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR