2-Ethyl-1,1'-biphenyl 98%

Supplier: Ambeed
Warning

A1004729-1G A1004729-250MG
76670-874EA 71.09 USD
76670-874 77619-524
2-Ethyl-1,1'-biphenyl 98%
2-Ethyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₄
MW: 182.2610 g/mol
MDL Number: MFCD30296301
CAS Number: 1812-51-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR