(4-(Cyclohexyloxy)phenyl)boronic acid 98%

Supplier: Ambeed
Warning

A1004722-10G A1004722-250MG A1004722-5G A1004722-1G
76670-868EA 214 USD
76670-868 76670-870 76670-872 77570-036
(4-(Cyclohexyloxy)phenyl)boronic acid 98%
(4-(Cyclohexyloxy)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₇BO₃
MDL Number: MFCD08701752
CAS Number: 570398-88-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR