2-(2,6-Dioxopiperidin-3-yl)-5-fluoroisoindoline-1,3-dione 97%
Supplier: Ambeed
|
Warning
|
A1004509-10G
A1004509-1G
A1004509-250MG
A1004509-25G
A1004509-5G
A1004509-100G
76670-856EA
77
USD
76670-856
76670-858
76670-860
76670-862
76670-864
77782-733
2-(2,6-Dioxopiperidin-3-yl)-5-fluoroisoindoline-1,3-dione 97%
2-(2,6-Dioxopiperidin-3-yl)-5-fluoroisoindoline-1,3-dione
Precaución: for Research Use Only
Formula:
C₁₃H₉FN₂O₄ |
MDL Number:
MFCD30188075 CAS Number: 835616-61-0 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...