Trimethyl(3-((trimethylsilyl)oxy)prop-1-yn-1-yl)silane 97%

Supplier: Ambeed
Danger

A1004019-1G A1004019-5G
76670-852EA 8.49 USD
76670-852 77570-034
Trimethyl(3-((trimethylsilyl)oxy)prop-1-yn-1-yl)silane 97%
Trimethyl(3-((trimethylsilyl)oxy)prop-1-yn-1-yl)silane

Precaución: for Research Use Only

Formula: C₉H₂₀OSi₂
MW: 200.4255 g/mol
CAS Number: 50965-66-7
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR