4-(Oxiran-2-ylmethoxy)butyl acrylate 97+% stabilized

Supplier: Ambeed
Danger


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A1003737-1G A1003737-5G A1003737-100G A1003737-25G
76670-848EA 9.55 USD
76670-848 76670-850 77570-028 77570-030
4-(Oxiran-2-ylmethoxy)butyl acrylate 97+% stabilized
4-(Oxiran-2-ylmethoxy)butyl acrylate

Precaución: for Research Use Only

Formula: C₁₀H₁₆O₄
MW: 200.2316 g/mol
MDL Number: MFCD30474906
CAS Number: 119692-59-0
UN: 3082
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR