Sulfamonomethoxine 95+%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-N-(6-methoxypyrimidin-4-yl)benzenesulphonamide, 4-Amino-N-(6-methoxy-4-pyrimidinyl)benzenesulphonamide, N-(6-Methoxy-4-pyrimidinyl)sulphanilamide

A1003464-100G A1003464-25G A1003464-500G A1003464-5G
76670-836EA 60.48 USD
76670-836 76670-838 76670-840 76670-842
Sulfamonomethoxine 95+%
Sulfamonomethoxine

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₄O₃S
MW: 280.30 g/mol
MDL Number: MFCD00063466
CAS Number: 1220-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR