Ethyl-2,4-dimethylnicotinate 96%

Supplier: Ambeed
Warning

A1003451-100G A1003451-10G A1003451-1G A1003451-25G A1003451-5G
76670-826EA 961 USD
76670-826 76670-828 76670-830 76670-832 76670-834
Ethyl-2,4-dimethylnicotinate 96%
Ethyl-2,4-dimethylnicotinate

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MDL Number: MFCD00051962
CAS Number: 37669-78-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR