Ethyl 1-cyclopentyl-5-hydroxy-1H-pyrazole-4-carboxylate 97%

Supplier: Ambeed
Warning

A1003419-100MG A1003419-1G A1003419-250MG
76670-820EA 36.08 USD
76670-820 76670-822 76670-824
Ethyl 1-cyclopentyl-5-hydroxy-1H-pyrazole-4-carboxylate 97%
Ethyl 1-cyclopentyl-5-hydroxy-1H-pyrazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₆N₂O₃
MW: 224.2563 g/mol
MDL Number: MFCD24344294
CAS Number: 1547888-86-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR