5-(Methylsulfonyl)nicotinonitrile 97%

Supplier: Ambeed
Warning

A1003161-100MG A1003161-250MG
76670-806EA 84.88 USD
76670-806 76670-808
5-(Methylsulfonyl)nicotinonitrile 97%
5-(Methylsulfonyl)nicotinonitrile

Precaución: for Research Use Only

Formula: C₇H₆N₂O₂S
MW: 182.1997 g/mol
MDL Number: MFCD30536550
CAS Number: 1780797-86-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR