D-(+)-Melezitose monohydrate 99+%

Supplier: Ambeed
Warning

Synonyms: (2R,2'R,3S,3'S,4S,4'S,5R,5'R,6R,6'R)-6,6'-(((2S,3S,4R,5R)-4-Hydroxy-2,5-bis(hydroxymethyl)tetrahydrofuran-2,3-diyl)bis(oxy))bis(2-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol) hydrate

A1002779-1G A1002779-25G A1002779-5G
76670-796EA 15 USD
76670-796 76670-798 76670-800
D-(+)-Melezitose monohydrate 99+%
D-(+)-Melezitose monohydrate

Precaución: for Research Use Only

Formula: C₁₈H₃₄O₁₇
MW: 522.453 g/mol
MDL Number: MFCD00149448
CAS Number: 10030-67-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR