1-Methyl-1,4-diazabicyclo[2.2.2]octan-1-ium iodide 97%

Supplier: Ambeed
Warning

A1002751-1G A1002751-5G
76670-792EA 28 USD
76670-792 77592-046
1-Methyl-1,4-diazabicyclo[2.2.2]octan-1-ium iodide 97%
1-Methyl-1,4-diazabicyclo[2.2.2]octan-1-ium iodide

Precaución: for Research Use Only

Formula: C₇H₁₅IN₂
MDL Number: MFCD30012657
CAS Number: 14968-74-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR