(S,S,S)-(3,5-Dioxa-4-phosphacyclohepta[2,1-a:3,4-a']dinaphthalen-4-yl)bis(1-phenylethyl)amine ≥97%, ee 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
(11bS)-N,N-Bis[(1S)-1-phenylethyl]dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-amine
A1002597-100MG
A1002597-1G
A1002597-5G
A1002597-250MG
76670-782EA
45.62
USD
76670-782
76670-784
76670-786
77619-488
(S,S,S)-(3,5-Dioxa-4-phosphacyclohepta[2,1-a:3,4-a']dinaphthalen-4-yl)bis(1-phenylethyl)amine ≥97%, ee 99%
(S,S,S)-(3,5-Dioxa-4-phosphacyclohepta[2,1-a:3,4-a']dinaphthalen-4-yl)bis(1-phenylethyl)amine
Precaución: for Research Use Only
Formula:
C₃₆H₃₀NO₂P MW: 539.61 g/mol |
MDL Number:
MFCD04117688 CAS Number: 209482-27-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...