Tetrahydro-2H-pyran-4-yl 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydro-1H-pyrrolo[3,2-b]pyridine-1-carboxylate 98%
Supplier: Ambeed
|
Warning
|
A1002593-100MG
A1002593-1G
A1002593-250MG
A1002593-5G
76670-774EA
128.37
USD
76670-774
76670-776
76670-778
76670-780
Tetrahydro-2H-pyran-4-yl 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydro-1H-pyrrolo[3,2-b]pyridine-1-carboxylate 98%
Tetrahydro-2H-pyran-4-yl 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydro-1H-pyrrolo[3,2-b]pyridine-1-carboxylate
Precaución: for Research Use Only
Formula:
C₁₉H₂₇BN₂O₅ MW: 374.2391 g/mol |
MDL Number:
MFCD33022343 CAS Number: 2828431-82-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...