Diethyl [2,2'-bipyridine]-3,3'-dicarboxylate 97%

Supplier: Ambeed
Warning

A1002579-100MG A1002579-1G A1002579-250MG A1002579-25G A1002579-5G
76670-764EA 12.74 USD
76670-764 76670-766 76670-768 76670-770 76670-772
Diethyl [2,2'-bipyridine]-3,3'-dicarboxylate 97%
Diethyl [2,2'-bipyridine]-3,3'-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₆H₁₆N₂O₄
MW: 300.3092 g/mol
MDL Number: MFCD06661494
CAS Number: 1762-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR