3-Bromo-2-chloro-5-cyanobenzoic acid 95%

Supplier: Ambeed
Warning

A1002109-100MG A1002109-1G A1002109-250MG
76670-746EA 88 USD
76670-746 76670-748 76670-750
3-Bromo-2-chloro-5-cyanobenzoic acid 95%
3-Bromo-2-chloro-5-cyanobenzoic acid

Precaución: for Research Use Only

Formula: C₈H₃BrClNO₂
MDL Number: MFCD28733663
CAS Number: 1805104-36-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR